accessibility menu, dialog, popup

UserName

CAS RN 186407-74-9

NHNBr

CAS RN 186407-74-9

IUPAC Name: 4-bromo-1H-indazole
Synonyms: 4-bromo-1H-indazole
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: KJIODOACRIRBPB-UHFFFAOYSA-N
SMILES: BrC1=C2C=NNC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results