CAS RN 186406-47-3
CAS RN 186406-47-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,2,4,4,6,6-Hexakis(2,2,3,3,4,4-hexafluorobutoxy)-2,2,4,4,6,6-hexahydro-1,3,5,2,4,6-Triazatriphosphorine, TRC
CAS: 186406-47-3 Molecular Formula: C24H18F36N3O6P3 Molecular Weight (g/mol): 1221.28 Synonym: Hexakis(1H,1H,4H-hexafluorobutyloxy)phosphazine; SMILES: FC(F)C(F)(F)C(F)(F)COP1(OCC(F)(F)C(F)(F)C(F)F)=NP(OCC(F)(F)C(F)(F)C(F)F)(OCC(F)(F)C(F)(F)C(F)F)=NP(OCC(F)(F)C(F)(F)C(F)F)(OCC(F)(F)C(F)(F)C(F)F)=N1