accessibility menu, dialog, popup

UserName

CAS RN 18638-99-8

H3CONH2OH3COH3C

CAS RN 18638-99-8

IUPAC Name: 1-(3,4,5-trimethoxyphenyl)methanamine
Synonyms: 1-(3,4,5-trimethoxyphenyl)methanamine
Molecular Weight (g/mol): 197.23
Molecular Formula: C10H15NO3
InChi Key: YUPUSBMJCFBHAP-UHFFFAOYSA-N
SMILES: COC1=CC(CN)=CC(OC)=C1OC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)