accessibility menu, dialog, popup

UserName

CAS RN 186306-70-7

H3COFFOH

CAS RN 186306-70-7

IUPAC Name: 2,3-difluoro-6-methoxyphenol
Synonyms: 2,3-difluoro-6-methoxyphenol
Molecular Weight (g/mol): 160.12
Molecular Formula: C7H6F2O2
InChi Key: HDQMXTCQIQIJFG-UHFFFAOYSA-N
SMILES: COC1=C(O)C(F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results