CAS RN 1863-63-4
CAS RN 1863-63-4
IUPAC Name:
ammonium benzoate
Synonyms:
ammonium benzoate
Molecular Weight (g/mol):
139.15
Molecular Formula:
C7H9NO2
InChi Key:
VWSRWGFGAAKTQG-UHFFFAOYSA-N
SMILES:
[NH4+].[O-]C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Ammonium benzoate, 99%
CAS: 1863-63-4 Molecular Formula: C7H9NO2 Molecular Weight (g/mol): 139.154 MDL Number: MFCD00013069 InChI Key: VWSRWGFGAAKTQG-UHFFFAOYSA-N Synonym: ammonium benzoate,benzoic acid, ammonium salt,ammonii benzoas,ammonium benzoicum,vulnoc ab,benzoic acid ammonium salt,hsdb 555,unii-ac80wd7gpf,benzoic acid, ammonium salt 1:1,ac80wd7gpf PubChem CID: 15830 IUPAC Name: azanium;benzoate SMILES: C1=CC=C(C=C1)C(=O)[O-].[NH4+]