accessibility menu, dialog, popup

UserName

HClOHNH

CAS RN 18621-18-6

IUPAC Name: hydrogen azetidin-3-ol chloride
Synonyms: hydrogen azetidin-3-ol chloride
Molecular Weight (g/mol): 109.55
Molecular Formula: C3H8ClNO
InChi Key: UQUPQEUNHVVNKW-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC1CNC1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)