accessibility menu, dialog, popup

UserName

FFBrFFH2C

CAS RN 18599-22-9

IUPAC Name: 4-bromo-3,3,4,4-tetrafluorobut-1-ene
Synonyms: 4-bromo-3,3,4,4-tetrafluorobut-1-ene
Molecular Weight (g/mol): 206.97
Molecular Formula: C4H3BrF4
InChi Key: GVCWGFZDSIWLMO-UHFFFAOYSA-N
SMILES: FC(F)(Br)C(F)(F)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)