accessibility menu, dialog, popup

UserName

H3C(S)CH3(S)NH2OOCH3HCl

CAS RN 18598-74-8

IUPAC Name: methyl (2S,3S)-2-amino-3-methylpentanoate
Synonyms: methyl (2S,3S)-2-amino-3-methylpentanoate
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: YXMMTUJDQTVJEN-WDSKDSINSA-N
SMILES: CC[C@H](C)[C@H](N)C(=O)OC
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)