accessibility menu, dialog, popup

UserName

CAS RN 18592-13-7

ClONHOHN

CAS RN 18592-13-7

IUPAC Name: 6-(chloromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 6-(chloromethyl)-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol): 160.56
Molecular Formula: C5H5ClN2O2
InChi Key: VCFXBAPEXBTNEA-UHFFFAOYSA-N
SMILES: ClCC1=CC(=O)NC(=O)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results