CAS RN 1851-09-8
IUPAC Name:
2-(4-chlorobenzenesulfonyl)acetonitrile
Synonyms:
2-(4-chlorobenzenesulfonyl)acetonitrile
Molecular Weight (g/mol):
215.65
Molecular Formula:
C8H6ClNO2S
InChi Key:
HAQGVGPNKGGSMK-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)S(=O)(=O)CC#N
1
–
1
of
1
results
1
(4-Chlorophenylsulfonyl)acetonitrile, 98+%, Thermo Scientific™
CAS: 1851-09-8 Molecular Formula: C8H6ClNO2S Molecular Weight (g/mol): 215.651 MDL Number: MFCD00045626 InChI Key: HAQGVGPNKGGSMK-UHFFFAOYSA-N Synonym: 4-chlorophenylsulfonylacetonitrile,2-4-chlorophenyl sulfonyl acetonitrile,4-chlorophenyl cyanomethyl sulfone,2-4-chlorobenzenesulfonyl acetonitrile,4-chlorobenzenesulphonylacetonitrile,4-chlorobenzenesulfonyl acetonitrile,4-chlorobenzenesulphonyl acetonitrile,4-chlorophenyl sulfonyl acetonitrile,2-4-chlorophenyl sulfonylacetonitrile,4-chlorophenylsulfonyl acetonitrile PubChem CID: 735829 IUPAC Name: 2-(4-chlorophenyl)sulfonylacetonitrile SMILES: C1=CC(=CC=C1S(=O)(=O)CC#N)Cl