accessibility menu, dialog, popup

UserName

CAS RN 18489-25-3

NNHNN

CAS RN 18489-25-3

IUPAC Name: 5-benzyl-2H-1,2,3,4-tetrazole
Synonyms: 5-benzyl-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 160.18
Molecular Formula: C8H8N4
InChi Key: HHDRWGJJZGJSGZ-UHFFFAOYSA-N
SMILES: C(C1=NNN=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results