accessibility menu, dialog, popup

UserName

CAS RN 18437-58-6

CH3NH2N

CAS RN 18437-58-6

IUPAC Name: 2-methylpyridin-4-amine
Synonyms: 2-methylpyridin-4-amine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: GNCLPNMQEGMNTG-UHFFFAOYSA-N
SMILES: CC1=NC=CC(N)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)