accessibility menu, dialog, popup

UserName

CH3OOOOOOH3CH3CCH3

CAS RN 18422-53-2

IUPAC Name: 2,2,5',5'-tetramethyl-tetrahydro-2H-spiro[[1,3]dioxolo[4,5-c]pyran-6,2'-[1,4]dioxolan]-7-one
Synonyms: 2,2,5',5'-tetramethyl-dihydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-6,2'-[1,4]dioxolan]-7-one
Molecular Weight (g/mol): 258.27
Molecular Formula: C12H18O6
InChi Key: IVWWFWFVSWOTLP-UHFFFAOYNA-N
SMILES: CC1(C)OC2COC3(COC(C)(C)O3)C(=O)C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results