accessibility menu, dialog, popup

UserName

H3COOSH2N

CAS RN 184174-81-0

IUPAC Name: methyl 2-amino-4H,5H,6H,7H,8H-cyclohepta[b]thiophene-3-carboxylate
Synonyms: methyl 2-amino-4H,5H,6H,7H,8H-cyclohepta[b]thiophene-3-carboxylate
Molecular Weight (g/mol): 225.31
Molecular Formula: C11H15NO2S
InChi Key: BDPLKEMLVDUYRR-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)SC2=C1CCCCC2
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results