accessibility menu, dialog, popup

UserName

CAS RN 18409-21-7

H3COH(E)(E)

CAS RN 18409-21-7

IUPAC Name: (2E,4E)-deca-2,4-dien-1-ol
Synonyms: (2E,4E)-deca-2,4-dien-1-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: NUBWFSDCZULDCI-BLHCBFLLSA-N
SMILES: CCCCC\C=C\C=C\CO
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)