accessibility menu, dialog, popup

UserName

CAS RN 1837-57-6

IUPAC Name: 2-hydroxypropanoic acid; 7-ethoxyacridine-3,9-diamine
Synonyms: ethacridine lactate
Molecular Weight (g/mol): 343.38
Molecular Formula: C18H21N3O4
InChi Key: IYLLULUTZPKQBW-UHFFFAOYNA-N
SMILES: CC(O)C(O)=O.CCOC1=CC=C2N=C3C=C(N)C=CC3=C(N)C2=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results