accessibility menu, dialog, popup

UserName

ClO(Z)

CAS RN 18365-42-9

IUPAC Name: (2Z)-2-chloro-3-phenylprop-2-enal
Synonyms: (2Z)-2-chloro-3-phenylprop-2-enal
Molecular Weight (g/mol): 166.6
Molecular Formula: C9H7ClO
InChi Key: SARRRAKOHPKFBW-TWGQIWQCSA-N
SMILES: Cl\C(C=O)=C/C1=CC=CC=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results