accessibility menu, dialog, popup

UserName

CAS RN 183428-90-2

CH3NH2NN

CAS RN 183428-90-2

IUPAC Name: 6-amino-2-methylpyridine-3-carbonitrile
Synonyms: 6-amino-2-methylpyridine-3-carbonitrile
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: OZAIJUZOQOBQDW-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(N)=N1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results