accessibility menu, dialog, popup

UserName

CAS RN 183365-31-3

NNH2

CAS RN 183365-31-3

IUPAC Name: 3-[(pyrrolidin-1-yl)methyl]aniline
Synonyms: 3-(pyrrolidin-1-ylmethyl)aniline
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: RZDBCURLSQNEAD-UHFFFAOYSA-N
SMILES: NC1=CC=CC(CN2CCCC2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results