CAS RN 183232-66-8
CAS RN 183232-66-8
IUPAC Name:
1-(2,4-dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-(piperidin-1-yl)-1H-pyrazole-3-carboxamide
Synonyms:
1-(2,4-dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-(piperidin-1-yl)pyrazole-3-carboxamide
Molecular Weight (g/mol):
555.24
Molecular Formula:
C22H21Cl2IN4O
InChi Key:
BUZAJRPLUGXRAB-UHFFFAOYSA-N
SMILES:
CC1=C(N(N=C1C(=O)NN1CCCCC1)C1=CC=C(Cl)C=C1Cl)C1=CC=C(I)C=C1
1
–
1
of
1
results
1
AM251, MedChemExpress
MedChemExpress AM251 is a selective cannabinoid 1 (CB1) receptor antagonist with an IC50 of 8 nM. AM251 also acts as a potent GPR55 agonist with an EC50 of 39 nM.