accessibility menu, dialog, popup

UserName

CAS RN 183208-35-7

IUPAC Name: 5-bromo-1H-pyrrolo[2,3-b]pyridine
Synonyms: 5-bromo-1H-pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: LPTVWZSQAIDCEB-UHFFFAOYSA-N
SMILES: BrC1=CN=C2NC=CC2=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)