accessibility menu, dialog, popup

UserName

CAS RN 183-97-1

IUPAC Name: 1,4,9,12-tetraoxadispiro[4.2.4⁸.2⁵]tetradecane
Synonyms: 1,4,9,12-tetraoxadispiro[4.2.4⁸.2⁵]tetradecane
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: YSMVSEYPOBXSOK-UHFFFAOYSA-N
SMILES: C1COC2(CCC3(CC2)OCCO3)O1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results