accessibility menu, dialog, popup

UserName

CAS RN 18297-63-7

H3CSiCH3H3CNHONHSiCH3CH3H3C

CAS RN 18297-63-7

IUPAC Name: 1,3-bis(trimethylsilyl)urea
Synonyms: 1,3-bis(trimethylsilyl)urea
Molecular Weight (g/mol): 204.42
Molecular Formula: C7H20N2OSi2
InChi Key: MASDFXZJIDNRTR-UHFFFAOYSA-N
SMILES: C[Si](C)(C)NC(=O)N[Si](C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results