accessibility menu, dialog, popup

UserName

CAS RN 182963-94-6

H3CNCH3ONH2

CAS RN 182963-94-6

IUPAC Name: 1-{3-[3-(dimethylamino)propoxy]phenyl}methanamine
Synonyms: 1-{3-[3-(dimethylamino)propoxy]phenyl}methanamine
Molecular Weight (g/mol): 208.31
Molecular Formula: C12H20N2O
InChi Key: VRMIFLYRNDVHIN-UHFFFAOYSA-N
SMILES: CN(C)CCCOC1=CC=CC(CN)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results