accessibility menu, dialog, popup

UserName

CAS RN 1825-61-2

H3COSiCH3CH3H3C

CAS RN 1825-61-2

IUPAC Name: methoxytrimethylsilane
Synonyms: methoxytrimethylsilane
Molecular Weight (g/mol): 104.22
Molecular Formula: C4H12OSi
InChi Key: POPACFLNWGUDSR-UHFFFAOYSA-N
SMILES: CO[Si](C)(C)C
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (6)
  • (3)