CAS RN 1824-52-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Bemetizide, TRC
CAS: 1824-52-8 Molecular Formula: C15 H16 Cl N3 O4 S2 Molecular Weight (g/mol): 401.89 Synonym: 6-Chloro-3,4-dihydro-3-(1-phenylethyl)-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-Dioxide,2H-1,2,4-Benzothiadiazine-7-sulfonamide, 6-chloro-3,4-dihydro-3-(alpha-methylbenzyl)-, 1,1-dioxide (6CI,7CI,8CI),Bemeticide,Bemetizide,Dehydrosanol,Diu 60,Su 7078 IUPAC Name: 6-chloro-1,1-dioxo-3-(1-phenylethyl)-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide SMILES: CC(C1Nc2cc(Cl)c(cc2S(=O)(=O)N1)S(=O)(=O)N)c3ccccc3