CAS RN 182362-38-5
CAS RN 182362-38-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
C26-Dihydro Ceramide, TRC
CAS: 182362-38-5 Molecular Formula: C44H89NO3 Molecular Weight (g/mol): 680.18 Synonym: [R-(R*,S*)]-N-[2-Hydroxy-1-(hydroxymethyl)heptadecyl]hexacosanamide,C26-Dihydroceramide,C26-Dihydro Ceramide IUPAC Name: N-[(2S,3R)-1,3-dihydroxyoctadecan-2-yl]hexacosanamide SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)N[C@@H](CO)[C@H](O)CCCCCCCCCCCCCCC