accessibility menu, dialog, popup

UserName

CH3NBrBrNHSH2N

CAS RN 1823514-27-7

IUPAC Name: (3,5-dibromo-6-methylpyridin-2-yl)thiourea
Synonyms: 3,5-dibromo-6-methylpyridin-2-ylthiourea
Molecular Weight (g/mol): 325.02
Molecular Formula: C7H7Br2N3S
InChi Key: RBDAYGUNTZANDA-UHFFFAOYSA-N
SMILES: CC1=NC(NC(N)=S)=C(Br)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results