accessibility menu, dialog, popup

UserName

BFOHHO

CAS RN 182344-25-8

IUPAC Name: (4-fluoronaphthalen-1-yl)boronic acid
Synonyms: 4-fluoronaphthalen-1-ylboronic acid
Molecular Weight (g/mol): 189.98
Molecular Formula: C10H8BFO2
InChi Key: IDCGAEJGZLYEAZ-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CC=CC2=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results