accessibility menu, dialog, popup

UserName

CH3N

CAS RN 1823-91-2

IUPAC Name: 2-phenylpropanenitrile
Synonyms: benzeneacetonitrile, α-methyl-
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: NVAOLENBKNECGF-UHFFFAOYNA-N
SMILES: CC(C#N)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)