accessibility menu, dialog, popup

UserName

OONClN

CAS RN 182181-19-7

IUPAC Name: 6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Synonyms: 6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Molecular Weight (g/mol): 195.58
Molecular Formula: C8H4ClN2O2
InChi Key: YZBKAIKDSQEETA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CN2C=C(Cl)C=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results