CAS RN 18174-72-6
CAS RN 18174-72-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Muscimol Hydrobromide, TRC
CAS: 18174-72-6 Molecular Formula: C4 H6 N2 O2 . Br H Molecular Weight (g/mol): 195.01 Synonym: Muscimol HBr IUPAC Name: 5-(aminomethyl)-1,2-oxazol-3-ol;hydrobromide SMILES: Br.NCc1onc(O)c1