accessibility menu, dialog, popup

UserName

CAS RN 17997-47-6

IUPAC Name: 2-(tributylstannyl)pyridine
Synonyms: 2-(tributylstannyl)pyridine
Molecular Weight (g/mol): 368.15
Molecular Formula: C17H31NSn
InChi Key: GYUURHMITDQTRU-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CC=CC=N1
Grade 
  • (4)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)