accessibility menu, dialog, popup

UserName

H3COFBOHHO

CAS RN 179897-94-0

IUPAC Name: (5-fluoro-2-methoxyphenyl)boronic acid
Synonyms: 5-fluoro-2-methoxyphenylboronic acid
Molecular Weight (g/mol): 169.95
Molecular Formula: C7H8BFO3
InChi Key: CCQKIRUMTHHPSX-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C=C1B(O)O
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)