CAS RN 179321-49-4
IUPAC Name:
tert-butyl N-(4-oxocyclohexyl)carbamate
Synonyms:
tert-butyl N-(4-oxocyclohexyl)carbamate
Molecular Weight (g/mol):
213.28
Molecular Formula:
C11H19NO3
InChi Key:
WYVFPGFWUKBXPZ-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NC1CCC(=O)CC1
1
–
1
of
1
results
1
4-(N-Boc-amino)cyclohexanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.