accessibility menu, dialog, popup

UserName

CAS RN 1791-13-5

H3CH3CCH3OOOOCH3H3CCH3H2NHCl

CAS RN 1791-13-5

IUPAC Name: 1,4-di-tert-butyl 2-aminobutanedioate hydrochloride
Synonyms: 1,4-di-tert-butyl 2-aminobutanedioate hydrochloride
Molecular Weight (g/mol): 281.78
Molecular Formula: C12H24ClNO4
InChi Key: GVLZIMQSYQDAHB-UHFFFAOYNA-N
SMILES: Cl.CC(C)(C)OC(=O)CC(N)C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)