accessibility menu, dialog, popup

UserName

CAS RN 179055-95-9

CH3NNN

CAS RN 179055-95-9

IUPAC Name: 3-(1-methyl-1H-pyrazol-5-yl)benzonitrile
Synonyms: 3-(2-methylpyrazol-3-yl)benzonitrile
Molecular Weight (g/mol): 183.21
Molecular Formula: C11H9N3
InChi Key: NEFOVTAGRMOIIV-UHFFFAOYSA-N
SMILES: CN1N=CC=C1C1=CC=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results