accessibility menu, dialog, popup

UserName

CAS RN 17846-68-3

IUPAC Name: azidotributylstannane
Synonyms: tributyltin azide
Molecular Weight (g/mol): 334.1
Molecular Formula: C12H29N3Sn
InChi Key: VJJINBNRYCPOOA-UHFFFAOYSA-N
SMILES: N=[N+]=[N-].CCCC[SnH](CCCC)CCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results