CAS RN 17834-04-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
8-Hydroxywarfarin, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
8-Hydroxy Warfarin, TRC
CAS: 17834-04-7 Molecular Formula: C19 H16 O5 Molecular Weight (g/mol): 324.33 Synonym: 2H-1-Benzopyran-2-one, 4,8-dihydroxy-3-(3-oxo-1-phenylbutyl)-,Coumarin, 3-(α-acetonylbenzyl)-4,8-dihydroxy- (8CI),8-Hydroxywarfarin,4,8-Dihydroxy-3-(3-oxo-1-phenylbutyl)-2H-1-benzopyran-2-one,8-Hydroxy warfarin IUPAC Name: 4,8-dihydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one SMILES: CC(=O)CC(C1=C(O)c2cccc(O)c2OC1=O)c3ccccc3