accessibility menu, dialog, popup

UserName

CAS RN 1780-36-5

CH3ClNClNCl

CAS RN 1780-36-5

IUPAC Name: 2,4,6-trichloro-5-methylpyrimidine
Synonyms: 2,4,6-trichloro-5-methylpyrimidine
Molecular Weight (g/mol): 197.44
Molecular Formula: C5H3Cl3N2
InChi Key: VTSWSQGDJQFXHB-UHFFFAOYSA-N
SMILES: CC1=C(Cl)N=C(Cl)N=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results