accessibility menu, dialog, popup

UserName

CAS RN 1779-25-5

H3CCH3AlClCH3CH3

CAS RN 1779-25-5

IUPAC Name: chlorobis(2-methylpropyl)alumane
Synonyms: diisobutylaluminium chloride
Molecular Weight (g/mol): 176.66
Molecular Formula: C8H18AlCl
InChi Key: HQMRIBYCTLBDAK-UHFFFAOYSA-M
SMILES: CC(C)C[Al](Cl)CC(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)