accessibility menu, dialog, popup

UserName

CAS RN 177760-52-0

CH3OOOH2NN

CAS RN 177760-52-0

IUPAC Name: ethyl 2-amino-1,3-oxazole-4-carboxylate
Synonyms: ethyl 2-amino-1,3-oxazole-4-carboxylate
Molecular Weight (g/mol): 156.14
Molecular Formula: C6H8N2O3
InChi Key: NBABLVASYFPOEV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=COC(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results