CAS RN 177735-09-0
CAS RN 177735-09-0
IUPAC Name:
(3-methylthiophen-2-yl)boronic acid
Synonyms:
3-methylthiophen-2-ylboronic acid
Molecular Weight (g/mol):
141.98
Molecular Formula:
C5H7BO2S
InChi Key:
MOOOGTOEKZWEJA-UHFFFAOYSA-N
SMILES:
CC1=C(SC=C1)B(O)O
1
–
1
of
1
results
1
3-Methylthiophene-2-boronic acid, 98%
CAS: 177735-09-0 Molecular Formula: C5H7BO2S Molecular Weight (g/mol): 141.979 MDL Number: MFCD00236049 InChI Key: MOOOGTOEKZWEJA-UHFFFAOYSA-N Synonym: 3-methylthiophene-2-boronic acid,3-methylthiophen-2-yl boronic acid,3-methyl-2-thienylboronic acid,3-methylthiophen-2-yl-2-boronic acid,3-methyl-2-thiopheneboronic acid,3-methyl-2-thienyl boronic acid,3-methyl-thiophen-2-boronic acid,3-methylthiophene-2-boronicacid,acmc-209ede,boronic acid, 3-methyl-2-thienyl PubChem CID: 3276437 IUPAC Name: (3-methylthiophen-2-yl)boronic acid SMILES: B(C1=C(C=CS1)C)(O)O