accessibility menu, dialog, popup

UserName

CAS RN 1777-03-3

CH3SiH3CCH3HC

CAS RN 1777-03-3

IUPAC Name: triethyl(ethynyl)silane
Synonyms: triethyl(ethynyl)silane
Molecular Weight (g/mol): 140.3
Molecular Formula: C8H16Si
InChi Key: FWSPXZXVNVQHIF-UHFFFAOYSA-N
SMILES: CC[Si](CC)(CC)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results