accessibility menu, dialog, popup

UserName

CAS RN 17766-26-6

NaNaNaSNHSNSHN

CAS RN 17766-26-6

IUPAC Name: trisodium 2,4,6-trisulfanylidene-1,3,5-triazinan-1-ide
Synonyms: trisodium 2,4,6-trisulfanylidene-1,3,5-triazinan-1-ide
Molecular Weight (g/mol): 245.22
Molecular Formula: C3H2N3Na3S3
InChi Key: NYVOYAFUSJONHU-UHFFFAOYSA-M
SMILES: [Na+].[Na+].[Na+].S=C1NC(=S)[N-]C(=S)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)