accessibility menu, dialog, popup

UserName

CAS RN 177492-52-3

IUPAC Name: 1,3-benzoxazol-6-amine
Synonyms: 1,3-benzoxazol-6-amine
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: ZJYIRVSPPOOPCL-UHFFFAOYSA-N
SMILES: NC1=CC=C2N=COC2=C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results