accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CCH3

CAS RN 177035-82-4

IUPAC Name: 2-dodecyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-dodecyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 296.3
Molecular Formula: C18H37BO2
InChi Key: WHOMEQHZQLKAPC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCB1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results