accessibility menu, dialog, popup

UserName

CAS RN 176976-42-4

HOBOHClFFF

CAS RN 176976-42-4

IUPAC Name: [4-chloro-3-(trifluoromethyl)phenyl]boronic acid
Synonyms: 4-chloro-3-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 224.37
Molecular Formula: C7H5BClF3O2
InChi Key: XHKNQBNGLMOTDB-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Cl)C(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results