accessibility menu, dialog, popup

UserName

CAS RN 17696-62-7

OHOO

CAS RN 17696-62-7

IUPAC Name: phenyl 4-hydroxybenzoate
Synonyms: phenylparaben
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: GJLNWLVPAHNBQN-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)OC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results