accessibility menu, dialog, popup

UserName

CAS RN 17692-51-2

H3CN(R)(R)(S)HNOONHCH3

CAS RN 17692-51-2

IUPAC Name: (2R,4S,7R)-4-({[(benzyloxy)carbonyl]amino}methyl)-6,11-dimethyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraen-6-ium
Synonyms: metergoline
Molecular Weight (g/mol): 404.53
Molecular Formula: C25H30N3O2
InChi Key: WZHJKEUHNJHDLS-QTGUNEKASA-O
SMILES: CN1C=C2C[C@@H]3[C@H](C[C@@H](CNC(=O)OCC4=CC=CC=C4)C[NH+]3C)C3=C2C1=CC=C3
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)